Orders LinkContactUsLinkComponent
Entomological forceps
Entomological forceps
Curved, wide tips, 120 mm
Catalog # 232-0038
Supplier:   MURRAY S
Entomological forceps
Catalog # 232-0038
Supplier:  MURRAY S
Entomological forceps
Entomological forceps
Curved, wide tips, 120 mm
Price
14.70 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel. Forceps for the collection of entomological specimens.
  • Guide pin helps to keep the tips aligned
  • Wide tips help to avoid damages of the specimen
  • Curved tips won't crush delicate larvae or eggs
Specifications
  • Type:
    Curved, wide tips
  • Length:
    120 mm