Orders LinkContactUsLinkComponent
Support clips
Support clips
Supplier:   BOCHEM
Support clips
Supplier:  BOCHEM
Certificates
Support clips
16 Options Available Below
Product Details & Documents
These support clips are coated with ceramic inside.
  • For tubes and cylindrical vessels
  • With fitting
Ordering information: The shafts (rods) need to be ordered separately.
Product Family Options
Shafts
Product Information
  • Availability
    Price
  • BOCH580
    Shaft, M12, for support clips, length 150 mm, O. Ø 15 mm
    Shaft, M12, for support clips, length 150 mm, O. Ø 15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 47.20
    SFr. 47.20
    1 * 1 ST
       SFr. 47.20
  • BOCH582
    Shaft, M12, for support clips, length 300 mm, O. Ø 15 mm
    Shaft, M12, for support clips, length 300 mm, O. Ø 15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 56.50
    SFr. 56.50
    1 * 1 ST
       SFr. 56.50
  • BOCH583
    Shaft, M12, for support clips, length 350 mm, O. Ø 15 mm
    Shaft, M12, for support clips, length 350 mm, O. Ø 15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 61.20
    SFr. 61.20
    1 * 1 ST
       SFr. 61.20
  • BOCH584
    Shaft, M12, for support clips, length 400 mm, O. Ø 15 mm
    Shaft, M12, for support clips, length 400 mm, O. Ø 15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 65.40
    SFr. 65.40
    1 * 1 ST
       SFr. 65.40
  • BOCH581
    Shaft, M12, for support clips, length 250 mm, O. Ø 15 mm
    Shaft, M12, for support clips, length 250 mm, O. Ø 15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 52.20
    SFr. 52.20
    1 * 1 ST
       SFr. 52.20
Product Information
  • Availability
    Price
  • BOCH590
    Support clip with fitting, O. Ø 25 - 28 mm
    Support clip with fitting, O. Ø 25 - 28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 88.40
    SFr. 88.40
    1 * 1 ST
       SFr. 88.40
  • BOCH591
    Support clip with fitting, O. Ø 31 - 35 mm
    Support clip with fitting, O. Ø 31 - 35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 89.20
    SFr. 89.20
    1 * 1 ST
       SFr. 89.20
  • BOCH592
    Support clip with fitting, O. Ø 40 - 44 mm
    Support clip with fitting, O. Ø 40 - 44 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 91.60
    SFr. 91.60
    1 * 1 ST
       SFr. 91.60
  • BOCH593
    Support clip with fitting, O. Ø 54 - 58 mm
    Support clip with fitting, O. Ø 54 - 58 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 104.00
    SFr. 104.00
    1 * 1 ST
       SFr. 104.00
  • BOCH594
    Support clip with fitting, O. Ø 58 - 61 mm
    Support clip with fitting, O. Ø 58 - 61 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 104.00
    SFr. 104.00
    1 * 1 ST
       SFr. 104.00
  • BOCH595
    Support clip with fitting, O. Ø 73 - 78 mm
    Support clip with fitting, O. Ø 73 - 78 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 119.00
    SFr. 119.00
    1 * 1 ST
       SFr. 119.00
  • BOCH596
    Support clip with fitting, O. Ø 85 - 90 mm
    Support clip with fitting, O. Ø 85 - 90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 141.00
    SFr. 141.00
    1 * 1 ST
       SFr. 141.00
  • BOCH597
    Support clip with fitting, O. Ø 100 - 104 mm
    Support clip with fitting, O. Ø 100 - 104 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 160.00
    SFr. 160.00
    1 * 1 ST
       SFr. 160.00
  • BOCH598
    Support clip with fitting, O. Ø 108 - 115 mm
    Support clip with fitting, O. Ø 108 - 115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 164.00
    SFr. 164.00
    1 * 1 ST
       SFr. 164.00
  • BOCH599
    Support clip with fitting, O. Ø 125 - 130 mm
    Support clip with fitting, O. Ø 125 - 130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 170.00
    SFr. 170.00
    1 * 1 ST
       SFr. 170.00
  • BOCH600
    Support clip with fitting, O. Ø 133 - 140 mm
    Support clip with fitting, O. Ø 133 - 140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 176.00
    SFr. 176.00
    1 * 1 ST
       SFr. 176.00