ContactUsLinkComponent
Counting chamber, Schilling-Kreuznetz
Counting chamber, Schilling-Kreuznetz
Catalog # HECH40452702
undefined
Counting chamber, Schilling-Kreuznetz
Catalog # HECH40452702
Specifications
  • Depth of chamber:
    0,1 mm
  • Type:
    Schilling-Kreuznetz
  • Description:
    Counting chamber
  • Pk:
    1
Specifications
About this item
Double lining counting chamber according to Schilling-Kreuznetz.