Filter Holders, Stainless Steel
: Merck
Stainless steel filter holders used to vacuum filter liquids for bacteriological or particulate contamination analysis.
Outlet: No. 8 perforated silicone stopper mounts in standard one liter filtering flask.
- Catalog No:
- MILFXX3001240
- MILFXF2004725
- MILFXF2004710
- Filtration area:
- 9 cm²
- 9 cm²
- Dimensions:
- Height: 19,4 mm
Ø: 7,6 mm - Height: 15,6 mm
Ø: 6,2 mm
Frequently Bought Together