Orders LinkContactUsLinkComponent
proteomics-chrom-955.jpg
Proteomics Chromatography Products
Innovative, reliable and easy-to-use, Thermo Scientific™ solutions help you meet the demanding needs at every stage of your proteomics workflow. From sample preparation to sample security and separation, we have the most comprehensive range of chromatography consumables.
Chromatography Consumables for Proteomics Workflows | Avantor
Product selection
CX1_400.jpg
Robust reproducible gradients that are applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.
Accucore_400.jpg
Based on Core Enhanced Technology, these columns provide fast resolution separation of peptides and proteins and generate only 50% of the back pressure associated with columns packed with sub 2 µm materials.
Acclaim_400.jpg
Achieve high-resolution peptide mapping and protein separations. The 3µm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase for fast, high-resolution separations.
SMART-digest-400.jpg
Highly stable immobilized trypsin reactor; when combined with state-of-the-art heating technology, it produces fast, reproducible digestions ready for analysis by LC/MS.
Nano_400.jpg
A comprehensive range of innovative columns designed to combine high performance with ease of use that deliver high performance separation to -omics research.