ContactUsLinkComponent
General Protection
Clothing that protects you in wide array of applications and environments from routine industrial to biopharmaceutical and aerospace. The ProShield® brand has broad offering to suit almost every requirement.
DuPont™-Tyvek1.jpg
DuPont™ Tyvek® 400
  • Composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont
  • Provides an ideal balance of protection, durability and comfort of any limited use fabric technology
  • Offers an inherent barrier against particles (down to 1.0 micron in size)
  • Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Learn more
DuPont™-Tyvek2.jpg
DuPont™ Tyvek® 400 D
  • Combines the protection, durability and comfort of DuPont™ Tyvek® fabric on the front and the comfort, softness and breathability of DuPont™ ProShield® 10 fabric on the back
  • Suitable for applications where comfort requirements are combined with limited protective requirements for frontal exposures
  • Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance
Learn more
Tyvek®-5003.jpg
DuPont™ Tyvek® 500
  • Composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont
  • Provides an ideal balance of protection, durability and comfort.
  • Available with external serged seams that provide the performance of Type 5/6 garments, which have been tested to standards against light liquid aerosols and airborne solid particles
  • Provides an effective barrier against particles (down to 1.0 micron in size)
  • Suitable for applications such as pharmaceutical handling, chemical processing, automatic spray painting, maintenance and many others.
Learn more
Tyvek®-6004.jpg
DuPont™ Tyvek® 600
  • Composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont
  • Provides an ideal balance of protection, durability and comfort
  • Available with serged and over-taped seams that provides the performance of Type 4/5/6 garments, which have been tested to standards against heavy liquid aerosols and airborne solid particles
  • Provides an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size)
  • Suitable for applications such as pharmaceutical manufacturing, medical applications, research and biosecurity laboratories, nuclear and maintenance
Learn more
Tyvek®-8005.jpg
DuPont™ Tyvek® 800
  • Combines resistance to low-concentration, water-based, inorganic chemicals (even under pressure) with the strength and durability of Tyvek® thanks to an innovative fabric technology and enhanced garment design
  • Available with serged and over-taped seams that provide the performance of Type 3/4/5/6 garments, which have been tested to standards against pressurized liquids, heavy liquid aerosols and airborne solid particles
  • Provides an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size), as well as oil repellency
  • Suitable for applications such as industrial cleaning, chemical packaging and redistribution, waste treatment and disposal, and environmental remediation
Learn more
ProShield6.jpg
DuPont™ ProShield® 10
  • Spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont
  • Optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments
  • Designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications
  • Available in a variety of coverall styles in a choice of white, blue and gray
  • Applications include: general maintenance, janitorial/cleaning and other dirty work
Learn more
ProShield507.jpg
DuPont™ ProShield® 50
  • Suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications
  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Applications include: janitorial, sanitation, and general industrial maintenance
Learn more
ProShield8.jpg
DuPont™ ProShield® 60
  • Suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications
  • Made with a microporous film that is laminated to a spundbonded polypropylene substrate
  • Applications include: automotive refinishing, waste cleanup and remediation
Learn more
BootCovers9.jpg
DuPont™ ProShield® 70 Shoe and Boot Covers
  • ProShield® 70 fabric, used exclusively for shoe and boot cover products, is a spunbonded polypropylene substrate that is coated with a low-density polyethylene film for enhanced skid resistance and barrier performance
Learn more