Orders LinkContactUsLinkComponent
UHPLC.jpg
Avantor® ACE® HPLC and UHPLC Columns
Avantor® manufacture the ultra-inert Avantor® ACE® UHPLC/HPLC column range. Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
Avantor® ACE® | Avantor
Avantor® ACE® LC phases have been specifically designed to help solve chromatographic challenges and include the unique C18-AR, C18-PFP, C18-Amide, CN-ES, SuperC18 and SuperPhenylHexyl phases, as well as a wide range of other 100 Å and 300 Å phases. Discover how Avantor® ACE® selectivities can help you with your chromatographic challenges. Avantor® ACE® - superior LC columns.
Avantor® ACE® Stationary Phase Portfolio
  • 17 different stationary phase selectivities based on fully porous inert silica
  • Additional 2 stationary phases based on solid core particle technology with extended pH range
  • 7 particle sizes with lengths between 30mm – 300mm
  • Choice of column id’s between 0.075mm – 30mm
Phase Guide
Avantor® ACE® Method Development Kits
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
See the products
Webinar
Brochure
Avantor® ACE® HILIC
Avantor® ACE® HILIC columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are available in three proprietary HILIC phases (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
Webinar
See the products
Brochure
Avantor® ACE® Method Development Kits with Chromsword® Method Development Software
Solve HPLC method development challenges systematically and efficiently – using Avantor® ACE columns and ChromSword 2 method development software. The Avantor® ACE ChromSword Method Development Kit includes 6 alternative selectivity Avantor® ACE phases, a copy of ChromSword 2 offline with a 6-month activation licence and a wealth of technical information and support – the perfect combination to introduce an improved method development protocol for your laboratory.
See the products
Webinar
Brochure
Avantor® ACE® Equivalence™ C18
Avantor® ACE® Equivalence C18 columns have been specifically developed as a high quality, cost effective replacement for many HPLC column brands.
  • Excellent efficiency and reproducibility
  • Available in 3 or 5 µm particle sizes
See the products
See the x-ref tool
Brochure
Avantor® ACE® Knowledge Zone
We are chromatographers dedicated to developing solutions that meet the demands of our fellow chromatographers working in pharmaceutical and biotechnology companies, universities, hospitals, research institutions, government agencies plus environmental and process control industries.
We have gained an unrivalled reputation for excellent quality and competitively priced products - backed up by expert technical support and after sales service.
In the Avantor® ACE® Knowledge Zone we are happy to share this knowledge with you.
Avantor® ACE® Knowledge Zone