Orders LinkContactUsLinkComponent
Set-0563-4401_New-feature.jpg
Meet testo 440 multifunction air velocity and IAQ measuring instrument
Meet your highest standards of air velocity and indoor-air-quality (IAQ) control
°C – - °F - CO2 – CO - %rF –%rH - fpm - m/s – hPa – Lux
More Information
Meet testo 440 multifunction air velocity and IAQ measuring instrument | Avantor
Imagine making a one-off investment in an air velocity and IAQ measuring instrument – and then being able to extend it easily and intuitive to suit your key applications.
One air velocity and IAQ measuring instrument supporting you reliable to meet your highest standards regarding quality assurance and safety regulations.
  • Just one instrument for all important air velocity and IAQ measurement demands
  • Intuitive wireless probe concept, quick and easy change of multiple digital probe heads
  • Highest product availability as digital probes can be calibrated without the instrument
  • Precise measurement of air flow in laboratory fume cupboards according to DIN EN 14175-3/-4
  • High accurate measurements especially for clean room applications
testo 440
Lab_Kit400-495.png
testo 440 air velocity & IAQ measuring instrument with internal memory and data export function. Comes with wired PT100 coated probe for aggressive media, and basic case for testo 440 and 1 probe.
Co2-KIT-BT400-495.png
testo 440 air velocity & IAQ measuring instrument, CO2 probe with Bluetooth, incl. temperature and humidity sensor, basic case for testo 440 and 1 probe.
Humidity-KIT400-495.png
testo 440 air velocity & IAQ measuring instrument, humidity and temperature probe with Bluetooth, basic case for testo 440 and 1 probe.
Lux-Kit400-495.png
testo 440 air velocity & IAQ measuring instrument, Lux probe, basic case for testo 440 and 1 probe.
Hot-wire-Kit400-495.png
testo 440 air velocity & IAQ measuring instrument with internal memory and data export function. Hot wire probe incl. temperature sensor, fixed cable (1.8 m) with telescope (0.85 m), basic case for testo 440 and 1 probe.
Product selection