ContactUsLinkComponent
READY PACK DOSYWEL UP! Distribution: Manual, automatic or with footswitch Adjustable volume: 0.1 to 9999 mL Adjustable speed: 5 - 100% / 10-450 rpm Calibration: At the beginning of a cycle and/or after a selected number of doses Dispensing modes: Dos 1 * 1 ST
Catalog # CMPPDOSW1201R
New Product
READY PACK DOSYWEL UP! Distribution: Manual, automatic or with footswitch Adjustable volume: 0.1 to 9999 mL Adjustable speed: 5 - 100% / 10-450 rpm Calibration: At the beginning of a cycle and/or after a selected number of doses Dispensing modes: Dos 1 * 1 ST
Catalog # CMPPDOSW1201R
New Product
Frequently Bought Together