Monkey Eotaxin ELISA Kit
Monkey Eotaxin ELISA Kit, 96 tests
Katalognr. ABCAAB317549-96TST
New Product
Anbieter: Abcam
Direkt zu
Product Details & Documents
Monkey Eotaxin ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey Eotaxin with a sensitivity of 12.913 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
Technische Daten
Technische Daten
- Bezeichnung:Monkey Eotaxin ELISA Kit
- Tests pro Kit:96 tests
- Empfindlichkeit:12,913 pg/ml
- Reaktivität der primären Antikörper:Monkey
- Testdauer:One step
- Time to Results:1 hr 30 min
- Cat. No.:ABCAAB317549-96TST
- Lagertemperatur:4 °C
- Format:Pre-coated
- Target protein:Eotaxin
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Serum, EDTA Plasma, Citrate plasma
- Erfassungsbereich:39,063 - 2500 pg/ml
- Regulatory status:RUO



