Human D-Dimer ELISA Kit
Human D-Dimer ELISA kit, 96 tests
Katalognr. ABCAAB315310-96TST
New Product
Anbieter: Abcam
Direkt zu
Product Details & Documents
Human D-Dimer ELISA kit is a single-wash 90-min Simplestep used to quantify Human D-Dimer with a sensitivity of 0,22 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
Technische Daten
Technische Daten
- Bezeichnung:Human D-Dimer ELISA kit
- Tests pro Kit:96 tests
- Empfindlichkeit:0,22 ng/ml
- Reaktivität der primären Antikörper:Human
- Testdauer:One step
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB315310-96TST
- Lagertemperatur:4 ℃
- Format:Pre-coated
- Target protein:D-Dimer
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Serum, EDTA Plasma, Citrate plasma, Heparin Plasma, Urine, Cell culture media
- Erfassungsbereich:0,781 - 50 ng/ml
- Regulatory status:RUO



