Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human D-Dimer ELISA Kit | Avantor

Human D-Dimer ELISA Kit

Human D-Dimer ELISA kit, 384 tests
Katalognr. ABCAAB315310-1X384
New Product
Anbieter:   Abcam
Human D-Dimer ELISA Kit
Katalognr. ABCAAB315310-1X384
Anbieter:  Abcam
New Product
CAS Nummer:  
Human D-Dimer ELISA Kit
Human D-Dimer ELISA Kit

Human D-Dimer ELISA kit, 384 tests

CAS Nummer:  
Preis
SFr. 1’930.00
Einloggen für Verfügbarkeit
1 * 1 Tests
Product Details & Documents

Human D-Dimer ELISA kit is a single-wash 90-min Simplestep used to quantify Human D-Dimer with a sensitivity of 0,22 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.

Technische Daten

Technische Daten

  • Bezeichnung:
    Human D-Dimer ELISA kit
  • Tests pro Kit:
    384 tests
  • Empfindlichkeit:
    0,22 ng/ml
  • Reaktivität der primären Antikörper:
    Human
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB315310-1X384
  • Lagertemperatur:
    4 ℃
  • Format:
    Pre-coated
  • Target protein:
    D-Dimer
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Serum, EDTA Plasma, Citrate plasma, Heparin Plasma, Urine, Cell culture media
  • Erfassungsbereich:
    0,781 - 50 ng/ml
  • Regulatory status:
    RUO

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.