Human IL-8 ELISA Kit
Product Details & Documents
Human IL-8 ELISA kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human interleukin-8 (IL-8) with a sensitivity of 1,8 pg/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for serum, plasma, cell culture supernatants, and cerebrospinal fluid - Cited in over 30 publications.
Specifications
Specifications
- Bezeichnung:Human IL-8 ELISA kit
- Tests pro Kit:384 tests
- Empfindlichkeit:1,8 pg/ml
- Reaktivität der primären Antikörper:Human
- Testdauer:One step
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB214030-1X384
- Lagertemperatur:4 °C
- Format:Pre-coated
- Target protein:IL-8
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Cerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
- Erfassungsbereich:3,91 - 250 pg/ml
- Regulatory status:RUO
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



