AuftragsverfolgungKontakt
CytoFLEX Mosaic Detection Modules | Avantor
CytoFLEX Mosaic Detection Modules
CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R (5 laser NUV)
Katalognr. BECMD20164
New Product
Anbieter:   Beckman Coulter
CytoFLEX Mosaic Detection Modules
Katalognr. BECMD20164
Anbieter:  Beckman Coulter
New Product
CytoFLEX Mosaic Detection Modules
CytoFLEX Mosaic Detection Modules
CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R (5 laser NUV)
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
  • A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful CytoFLEX flow cytometry solution to date.
Technische Daten
  • Bezeichnung:
    CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system N-V-B-Y-R (5 laser NUV)
  • Seq:
    11