4-(2,4-Dinitrophenyl)morpholine
Bottle
Katalognr. APOSOR200048-100MG
Anbieter: Apollo Scientific
4-(2,4-Dinitrophenyl)morpholine
Katalognr. APOSOR200048-100MG
Anbieter: Apollo Scientific
CAS Nummer: 39242-76-7
Direkt zu
Product Details & Documents
- CAS Nummer:39242-76-7
- Formel:C₁₀H₁₁N₃O₅
- MDL-Nummer:MFCD00193779
- Molekulargewicht:253.22 g/mol
- Schmelzender Platingehalt:117…118 °C
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



