Methylnonadecanoate 98%
Bottle
Katalognr. APOSOR1013903-5G
Anbieter: Apollo Scientific
Methylnonadecanoate 98%
Katalognr. APOSOR1013903-5G
Anbieter: Apollo Scientific
CAS Nummer: 1731-94-8
Direkt zu
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyme: Nonadecanoic acid methyll ester, Methyl Nonadecanoate
- CAS Nummer:1731-94-8
- Formel:C₂₀H₄₀O₂
- MDL-Nummer:MFCD00009011
- Molekulargewicht:312,53 g/mol
Technische Daten
Technische Daten
- Packung:Bottle



