Methyl Tridecanoate 98%
Bottle
Katalognr. APOSOR1013885-100G
Anbieter: Apollo Scientific
Methyl Tridecanoate 98%
Katalognr. APOSOR1013885-100G
Anbieter: Apollo Scientific
CAS Nummer: 1731-88-0
Direkt zu
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyme: Tridecylic acid methyl ester, Tridecanoic acid methyl ester
- CAS Nummer:1731-88-0
- Formel:C₁₄H₂₈O₂
- MDL-Nummer:MFCD00008977
- Molekulargewicht:228,37 g/mol
Technische Daten
Technische Daten
- Packung:Bottle



