Fluorescein 95%
Bottle
Katalognr. APOSOR1013317-500G
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Usage: Organic building block/reagent
Synonyme: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Nummer:2321-07-5
- Formel:C₂₀H₁₂O₅
- MDL-Nummer:MFCD00005050
- Molekulargewicht:332,31 g/mol
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



