2-(Methylthio)benzaldehyd 90%
Bottle
Katalognr. APOSOR938789-25G
Anbieter: Apollo Scientific
2-(Methylthio)benzaldehyd 90%
Katalognr. APOSOR938789-25G
Anbieter: Apollo Scientific
CAS Nummer: 7022-45-9
Direkt zu
Product Details & Documents
- CAS Nummer:7022-45-9
- Formel:C₈H₈OS
- MDL-Nummer:MFCD00196822
- Molekulargewicht:152.2135 g/mol
- Siedepunkt:141…143 °C
- Dichte:1,18 g/cm³ (25 °C)
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



