15 mL Vial Rack for Cleaning System Holds (36) 15 mL vials and closures PFA racks with PTFE post and handle Drip tray included Designed for use with vial cleaning system 550-01 May be used with some 4 L glass beakers (requires PFA beaker lid) 1 * 1 ST
Katalognr. SAVI550-15
Anbieter: SAVILLEX
New Product
Frequently Bought Together