Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface | Avantor
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
TableWorx performance work table, type 304 stainless steel work surface, legs, and leg mounts, stainless steel undershelf, model: TW-FS-304-S, 762×1524 mm
Katalognr. METOTWM3060FS304S
Anbieter:   Metro International
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
Katalognr. METOTWM3060FS304S
TableWorx Performance Work Tables, 304 Stainless Steel Work Surface
TableWorx performance work table, type 304 stainless steel work surface, legs, and leg mounts, stainless steel undershelf, model: TW-FS-304-S, 762×1524 mm
Preis
SFr. 3’620.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
TableWorx is a truly modular platform of performance work tables designed to allow you to customise your perfect solution.
  • Colour: Silver
All stainless series TableWorx performance work tables feature a 16-gauge, type 304 stainless steel work surface reinforced with 16-gauge hat channels. The sound deadened work surface features 90° turned down edges on all four sides. All sizes are available with an optional 51 mm stainless steel backsplash, which is formed into the work surface.
All stainless series TableWorx performance work tables feature type 304 stainless steel legs and leg mounts. A solid, type 304 stainless steel undershelf provides additional storage space.
Stationary work table legs include stainless steel leveling bullet feet with 29 mm range of adjustability. Mobile-ready versions are also available, and can accept any Metro 25 mm stem caster. Please note that casters are NOT included with mobile-ready TableWorx models and must be ordered separately.
Elevate your efficiency to new levels with the addition of TableWorx productivity riser. Risers can be easily added to any TableWorx work table. A full array of overshelves, wire grids, and accessories can be added to maximise every inch of space, making you more comfortable and productive than ever before. Risers can be mounted at the rear or center of the work table.
Technische Daten
  • Bezeichnung:
    TableWorx performance work table, type 304 stainless steel work surface, legs, and leg mounts, stainless steel undershelf, model: TW-FS-304-S, 762×1524 mm
  • Seq:
    11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.