SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
[EN]36 COMPONENTS: 200MG/L EACH 1 * 5 Ampull
CMPLRD0204911.5
:
CUSTOM MADE CHEMICALS LAB
New Product
[EN]36 COMPONENTS: 200MG/L EACH 1 * 5 Ampull
CMPLRD0204911.5
:
CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Peressigsäure-Teststäbchen, MQuant®, Supelco®
: Merck
Ammoniaklösung 25%, EMSURE® ISO, Reag. Ph. Eur. zur Analyse, Supelco®
: Merck
Natriumhydroxid 98,5-100,5%, Plätzchen, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
: VWR Chemicals
Teststreifen, Nitrat/Nitrit, Dosatest
: VWR Chemicals
Testpapiere pH, MQuant®, Supelco®
: Merck
Nitrat-Teststäbchen, MQuant®, Supelco®
: Merck
Ammoniaklösung 25%, AnalaR NORMAPUR® analytisches Reagens
: VWR Chemicals
Natriumhydroxid, Plätzchen, EMSURE® zur Analyse, Supelco®
: Merck
Testsätze, Phosphat, Spectroquant®, Supelco®
: Merck
Natriumchlorid, EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®
: Merck
Chlor-Teststäbchen, MQuant®, Supelco®
: Merck
Sulfit-Teststäbchen, MQuant®, Supelco®
: Merck
Testsätze, CSB-Küvettentest, Spectroquant®, Supelco®
: Merck
Peroxid-Teststäbchen, MQuant®, Supelco®
: Merck
Natriumhydroxid, Plätzchen, rein, Supelco®
: Merck
pH-Indikatorstäbchen, MQuant®, Supelco®
: Merck