L(-)-Prolin *copeia
Plastic bottle, 100 g
Katalognr. CMPP2031-100G
Anbieter: CUSTOM MADE CHEMICALS
Direkt zu
Product Details & Documents
Synonyme: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
- CAS Nummer:147-85-3
- Formel:C₅H₉NO₂
- MDL-Nummer:MFCD00064318
- Molekulargewicht:115.13 g/mol
- Lagertemperatur:Room Temperature
- EINECS:205-702-2
Technische Daten
Technische Daten
- Packung:Plastic bottle
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



