KOFLO CORPORATION 0.375 DIAMETER SINGLE-USE PHARMACEUTICAL STATIC MIXER - TWELVE (12) USP CLASS VI KYNAR PVDF HELICAL MIXING ELEMEN TS - FDA-REGISTERED ISO CLASS 7 CLEANROOM MANUFACTURED AND PACKAGED - 2 UNITS REQUIRED IN SERIES 1 * 1 ST
Catalog # MSPPSU037512H-PVDF
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together