Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]14 COMPONENTS IN HNO3 10% 1 * 3 Fl
Catalog # CMPLRM076381L25.3
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Expand image
[EN]14 COMPONENTS IN HNO3 10% 1 * 3 Fl
Catalog # CMPLRM076381L25.3
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
PHOSPHATE BUFFERED SALINE 1 * 10 L
Supplier: Avantor
Referenzsubstanzen
Supplier: Merck
Teststifte Oberflächenenergie 30-44 mN/m 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
NEXTAL PEGS SINGLE CONDITION REFILL 1 * 100 mL
Supplier: MARKET SOURCE PART PROCESS
[EN]ANTIPD1 RMP114 ULTRALOW ENDOTOXIN 5M 1 * 100 mg
Supplier: MARKET SOURCE PART PROCESS
Diluid 100 Plus für die Hämatologie (max. 0,001% Aceton), J.T.Baker®
Supplier: Avantor
VWR® Pulverpackungen zur Eisenbestimmung, Reagenzien für Photometer
Supplier: VWR Chemicals
C57BL/6 Mouse Cardiac Fibroblasts 1 * 1 Vial
Supplier: MARKET SOURCE PART PROCESS