[EN]1 COMPONENT: HG 0.5MG/L IN HCL 5% 1 * 250 mL
Katalognr. CMPLRM030240L25
Anbieter: CUSTOM MADE CHEMICALS LAB
New Product
[EN]1 COMPONENT: HG 0.5MG/L IN HCL 5% 1 * 250 mL
Katalognr. CMPLRM030240L25
Anbieter: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Ammoniaklösung 25%, AnalaR NORMAPUR® analytisches Reagens
Anbieter: VWR Chemicals
Testsätze, Phosphat, Spectroquant®, Supelco®
Anbieter: Merck
Nitrat-Teststäbchen, MQuant®, Supelco®
Anbieter: Merck
Teststreifen, Peressigsäure, Dosatest
Anbieter: VWR Chemicals
Testsätze, Ammonium, Spectroquant®, Supelco®
Anbieter: Merck
pH-Indikatorstäbchen, MQuant®, Supelco®
Anbieter: Merck
Testsätze, CSB-Küvettentest, Spectroquant®, Supelco®
Anbieter: Merck
Testsätze, Tenside, Spectroquant®, Supelco®
Anbieter: Merck
Natriumhydroxid 98,5-100,5%, Plätzchen, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Anbieter: VWR Chemicals
Testsätze, Nitrat, Spectroquant®, Supelco®
Anbieter: Merck
Supelco® Spectroquant® Testkits, Sulfat
Anbieter: Merck
Nitrit VHR L, 500 ml
Anbieter: Lovibond