Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry | Avantor

ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry

ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 100 mL
Katalognr. AATB22871
Anbieter:   AAT Bioquest
ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
Katalognr. AATB22871
Anbieter:  AAT Bioquest
CAS Nummer:  
ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry

ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 100 mL

CAS Nummer:  
Preis
143.00 SFr.
Einloggen für Verfügbarkeit
1 * 100 mL
Product Details & Documents

The ReadiUse™ Annexin Binding Buffer *Optimised for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.

  • Provided as a 10X concentrate
  • Saves time
  • pH of 7.4

Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.

Caution: For research use only.

Technische Daten

Technische Daten

  • VE:
    100 mL
  • Bezeichnung:
    ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry

Technische Daten