SPR SENSOR CHIP. BIACORE SERIES S COMPATIBLE COATING: 200 NM OLIGONUCLEOTIDE DERIVATIZED CARBOXYMETHLYDEXTRAN MEDIUM DENSITY. FOR RE VERSIBLE LIGAND IMMOBILIZATION. 1 * 1 ST
Katalognr. MSPPSCBSRGD200M
Anbieter: MARKET SOURCE PART PROCESS
Frequently Bought Together