AuftragsverfolgungKontakt
Bacillol® 30 Sensitive Green Tissues | Avantor
Bacillol® 30 Sensitive Green Tissues
Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces
Katalognr. BODE9819430_1
Anbieter:   BODE CHEMIE
Bacillol® 30 Sensitive Green Tissues
Katalognr. BODE9819430_1
Anbieter:  BODE CHEMIE
Bacillol® 30 Sensitive Green Tissues
Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces
Preis
SFr. 103.00
Einloggen für Verfügbarkeit
1 * 6 ST
Product Details & Documents
The ready-to-use Bacillol® 30 sensitive green tissues disinfectant wipes from the flowpack, soaked with Bacillol® 30 sensitive foam, wet the surfaces excellently and work quickly, even on sensitive surfaces.
  • 100% plastic-free wipes
  • Free of microplastics
  • Fast-acting with good cleaning properties
  • Particularly gentle on materials, also suitable for sensitive materials such as: polycarbonate (PC), acrylic glass (PMMA) and polysulfone
  • It can be used without gloves
  • Skin-friendly and allergy-friendly
Effectiveness Bactericidal, yeasticidal, tuberculocidal, mycobactericidal, limited virucidal (incl. HBV, HIV, HCV), limited virucidal PLUS.
Zertifizierung: ​​SGS Institut Fresenius**- and ECARF-certified.
Technische Daten
  • Bezeichnung:
    Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces