Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Bacillol® 30 Sensitive Green Tissues
Bacillol® 30 Sensitive Green Tissues
Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces
Codice catalogo BODE9819430
Nome del fornitore:   BODE CHEMIE
Bacillol® 30 Sensitive Green Tissues
Codice catalogo BODE9819430
Nome del fornitore:  BODE CHEMIE
Bacillol® 30 Sensitive Green Tissues
Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces
Price
17.10 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The ready-to-use Bacillol® 30 sensitive green tissues disinfectant wipes from the flowpack, soaked with Bacillol® 30 sensitive foam, wet the surfaces excellently and work quickly, even on sensitive surfaces.
  • 100% plastic-free wipes
  • Free of microplastics
  • Fast-acting with good cleaning properties
  • Particularly gentle on materials, also suitable for sensitive materials such as: polycarbonate (PC), acrylic glass (PMMA) and polysulfone
  • It can be used without gloves
  • Skin-friendly and allergy-friendly
Effectiveness Bactericidal, yeasticidal, tuberculocidal, mycobactericidal, limited virucidal (incl. HBV, HIV, HCV), limited virucidal PLUS.
Certifications: ​​SGS Institut Fresenius**- and ECARF-certified.
Specifiche
  • Bezeichnung:
    Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.