Sucralose 98%
Plastic bottle, 100 g
470091000.
: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside
- :56038-13-2
- :C₁₂H₁₉Cl₃O₈
- :MFCD03648615
- :397.63 g/mol
- :13,08965
- :Raumtemperatur
- :259-952-2
- Appearance:white to almost white crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:+84° to +88° (20°C, 589 nm) (C=1% in water) (on dry substance)
- Water:≤2 %
- Packung:Plastic bottle
- VE:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



