Sucralose 98%
Plastic bottle, 1 kg
470090010.
: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside
- :56038-13-2
- :C₁₂H₁₉Cl₃O₈
- :MFCD03648615
- :397.63 g/mol
- :13,08965
- :Raumtemperatur
- :259-952-2
- Appearance:white to almost white crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:+84° to +88° (20°C, 589 nm) (C=1% in water) (on dry substance)
- Water:≤2 %
- Packung:Plastic bottle
- VE:1 kg



