[EN]TBNG MFLEX PURI-FLEX I/P 73 150'/45, 1 * 4.572 cm
Katalognr. MFLX96419-73-S150
Anbieter: Avantor Fluid Handling
[EN]TBNG MFLEX PURI-FLEX I/P 73 150'/45, 1 * 4.572 cm
Katalognr. MFLX96419-73-S150
Anbieter: Avantor Fluid Handling
Frequently Bought Together
Schlauchklemmen, SP Bel-Art
Anbieter: Bel-Art Products, a Part of SP
Tubing
Anbieter: GILSON
Volumetric scoops, USP Class VI, double-bagged, ULTRA
Anbieter: Sampling Systems
Entlüftungsschlauch, b.safe
Anbieter: Bohlender
Biopharmaceutical tubing C-Flex® , Formulation 082
Anbieter: Saint Gobain Life Sciences
[EN]TBG PHARMED 1/50X9/64'' 25'/7.62M 1 * 25 Fuß
Anbieter: Avantor Fluid Handling
Masterflex® Ismatec® Pumpenschlauch, 2-Stopp-Microbore, PharMed® BPT, Avantor®
Anbieter: Avantor Fluid Handling
[EN]3/32 X 15/32 PT TBNG 1 * 25 ST
Anbieter: Avantor Fluid Handling
Schläuche, Silikon, K70
Anbieter: KARTELL
Cork rings for round bottom flasks
Anbieter: BRAEMSWIG
Scoops, USP Class VI, double-bagged, ULTRA
Anbieter: Sampling Systems
Plug Valves with Compression Fittings, PFA
Anbieter: Avantor Fluid Handling
Schlauchverbinder, gerade, zylindrische Tüllen, LaboPlast®
Anbieter: BURKLE
Tubing, Silicone, Sani-Tech®, ULTRA-C Series
Anbieter: Saint Gobain Life Sciences
Schläuche, Gummi
Anbieter: Roth Carl
Black Rubber Vacuum Transfer Pump Tubing
Anbieter: Avantor Fluid Handling
Accessories for SP Bel-Art Lab Companion™ Desiccator Cabinets
Anbieter: Bel-Art Products, a Part of SP