INFIGRTINIB FREE BASE 1 * 100 mg
ENZOENZCHM2250100
Frequently Bought Together
AlloSeq Tx17 (RUO) Set A 1 * 1 KIT
: MARKET SOURCE PART PROCESS
[EN]6 COMPONENTS IN HNO3 2% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
Kaliumhydroxid ≥85%, Schuppen
: Thermo Fisher Scientific
[EN]1 COMPONENT OS 1000MG/L IN HCL 2% 1 * 100 mL
: CUSTOM MADE CHEMICALS LAB
Powder pillows, sulphate, Sulfa 4
: Lovibond
N,O-Bis(trimethylsilyl)trifluoracetamid mit 1% Chlortrimethylsilan
: Thermo Fisher Scientific