Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Green detection kit, LYSO-ID®
Green detection kit, LYSO-ID®
Green detection kit, LYSO-ID®, 100 Tests
Catalog # ENZOENZ510340100
Supplier:   Enzo Life Sciences
Green detection kit, LYSO-ID®
Catalog # ENZOENZ510340100
Supplier:  Enzo Life Sciences
CAS Number:  
Green detection kit, LYSO-ID®
Green detection kit, LYSO-ID®, 100 Tests
CAS Number:  
Price
SFr. 202.00
Log in for availability
1 * 100 Tests
Product Details & Documents
LYSO-ID® Green Detection Kit is an acidic organelle-selective dye specially designed to use with RFP-expressing cell lines, as well as cells expressing blue, cyan or orange fluorescent proteins (BFPs, CFPs, OFPs).
  • Used for both short-term and long-term tracking studies
  • Low micromolar concentrations is sufficient for staining mammalian cells
  • Accumulates in acidic compartments (endosomes, lysosomes, and secretory vesicles)
  • Generates emission profiles that can be multiplexed with other fluorophores
LYSO-ID® Green dye is a new green-emitting, cell-permeable small organic probe molecule that spontaneously localizes to live cell acidic organelles. It can be readily used in combination with other common UV and visible light excitable fluorescent dyes and various fluorescent proteins in multicolor imaging and detection applications.
Delivery information: Kit contains LYSO-ID® Green Detection Reagent, Hoechst 33342 nuclear stain, Chloroquine control and 10X assay buffer.
Specifications
Specifications
  • VE:
    100 Tests
  • Bezeichnung:
    Green detection kit, LYSO-ID®
Specifications