Green detection kit, MITO-ID®
Product Details & Documents
MITO-ID® Green Detection Kit is a potent mitochondria-selective dye suitable for live, permeabilised and fixed cell staining.
- Easily multiplexed with common fluorescent dyes (coumarin, cyanine 5) and fluorescent proteins (BFP, CFP, RFP)
- Highly resistant to photobleaching and concentration quenching
- Ensures strong and consistent fluorescence signal, even after extended viewing periods
- Highlights mitochondria regardless of the organelle's membrane potential status
- Stringently manufactured, to control and eliminate non-specific assay artifacts
MITO-ID® Green Detection Kit is useful for assessing mitochondrial morphology changes, estimating mitochondrial mass and co-localising RFP-tagged proteins to the mitochondrial compartment. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes as well as High Content Screening (HCS) platforms.
: Kit contains MITO-ID® Green Detection Reagent, Hoechst 33342 nuclear stain and 10X assay buffer.
Technische Daten
Technische Daten
- VE:500 Tests
- Bezeichnung:Green detection kit, MITO-ID®