Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Green detection kit, MITO-ID®
Green detection kit, MITO-ID®
Green detection kit, MITO-ID®, 100 Tests
Katalognr. ENZOENZ510220100
Anbieter:   Enzo Life Sciences
Green detection kit, MITO-ID®
Katalognr. ENZOENZ510220100
Anbieter:  Enzo Life Sciences
CAS Nummer:  
Green detection kit, MITO-ID®
Green detection kit, MITO-ID®, 100 Tests
CAS Nummer:  
Preis
SFr. 152.00
Einloggen für Verfügbarkeit
1 * 100 Tests
Product Details & Documents
MITO-ID® Green Detection Kit is a potent mitochondria-selective dye suitable for live, permeabilised and fixed cell staining.
  • Easily multiplexed with common fluorescent dyes (coumarin, cyanine 5) and fluorescent proteins (BFP, CFP, RFP)
  • Highly resistant to photobleaching and concentration quenching
  • Ensures strong and consistent fluorescence signal, even after extended viewing periods
  • Highlights mitochondria regardless of the organelle's membrane potential status
  • Stringently manufactured, to control and eliminate non-specific assay artifacts
MITO-ID® Green Detection Kit is useful for assessing mitochondrial morphology changes, estimating mitochondrial mass and co-localising RFP-tagged proteins to the mitochondrial compartment. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes as well as High Content Screening (HCS) platforms.
Delivery information: Kit contains MITO-ID® Green Detection Reagent, Hoechst 33342 nuclear stain and 10X assay buffer.
Technische Daten
Technische Daten
  • VE:
    100 Tests
  • Bezeichnung:
    Green detection kit, MITO-ID®
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.