Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Cell counting kit-F | Avantor

Cell counting kit-F

Cell counting kit-F, 5×500, 500 Tests
Katalognr. ENZOALX850245KI02
Anbieter:   Enzo Life Sciences
Cell counting kit-F
Katalognr. ENZOALX850245KI02
Anbieter:  Enzo Life Sciences
CAS Nummer:  
Cell counting kit-F

Cell counting kit-F, 5×500, 500 Tests

CAS Nummer:  
Preis
SFr. 1’120.00
Einloggen für Verfügbarkeit
1 * 500 Tests
Product Details & Documents

Fluorometric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages includes no radioisotopes, more sensitive than the colorimetric assay, short staining reaction time, no solubilization step, correlates with the [3H]-thymidine incorporation assay.

Technische Daten

Technische Daten

  • VE:
    500 Tests
  • Bezeichnung:
    Cell counting kit-F
  • Tests pro Kit:
    5×500
  • Anwendung:
    Fluorescence Microscopy
  • Cat. No.:
    ENZOALX850245KI02
  • Größe:
    5×500 tests
  • Lagerbedingungen:
    −20 °C

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.