Serotonin Hydrochlorid
Glass bottle for solids, 50 mg
Katalognr. ENZOALX550328M050
Anbieter: Enzo Life Sciences
Serotonin Hydrochlorid
Katalognr. ENZOALX550328M050
Anbieter: Enzo Life Sciences
CAS Nummer: 153-98-0
Direkt zu
Product Details & Documents
Synonyme: 5-Hydroxytryptamine Hydrochloride
- CAS Nummer:153-98-0
- Formel:C₁₀H₁₃ClN₂O
- MDL-Nummer:MFCD00012686
- Molekulargewicht:212.68 g/mol
- UN:2811
- ADR:6.1,III
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:50 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



