Gentamicinsulfat
Glass bottle for solids, 5 g
ENZOALX380003G005
: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
- :1405-41-0
- :MFCD00270181
- :13,04403
- :218…237 °C
- :Raumtemperatur
- :215-778-9
- Packung:Glass bottle for solids
- VE:5 g