Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
[D-Asp3]microcystin-RR | Avantor

[D-Asp3]microcystin-RR

100 µG
Katalognr. ENZOALX350168C100
Anbieter:   Enzo Life Sciences
[D-Asp3]microcystin-RR
Katalognr. ENZOALX350168C100
Anbieter:  Enzo Life Sciences
CAS Nummer:  
[D-Asp3]microcystin-RR
[D-Asp3]microcystin-RR

100 µG

CAS Nummer:  
Preis
SFr. 707.00
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents

Microcystins are a group of cyclic heptapeptide hepatotoxins produced by a number of cyanobacterial genera. The most notable of which, and namesake, is the widespread genus Microcystis. Structurally, all microcystins consist of the generalized structure cyclo(-D-Ala1-X2-D-MeAsp3-Y4-Adda5-D-Glu6-Mdha7-). X and Y are variable L-amino acids, D-MeAsp is D-erythro-β-methylaspartic acid and Mdha is N-methyldehydroalanine. Adda is the cyanobacteria unique C20 β-amino acid 3-amino-9-methoxy-2,6,8-trimethyl-10-phenyl-deca-4,6-dienoic acid. Substitutions of the variable L-amino acids at positions 2 and 4 give rise to at least 21 known primary microcystin analogs and alterations in the other constituent amino acids result in more than 90 reported mycrocystins to date.

Isolated from Microcystis aeruginosa. Microcystin containing arginine (R) in positions 2 and 4. Additionally, the D-MeAsp at position 3 is demethylated to Asp. Hepatotoxic.

Technische Daten

Technische Daten

  • VE:
    100 µG
  • Cat. No.:
    ENZOALX350168C100
  • Purity:
    >95% (HPLC)
  • Protein/Peptid-Name:
    [D-Asp3]microcystin-RR

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.