Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Microcystin-LR
Microcystin-LR
100 µG
Katalognr. ENZOALX350012C100
Anbieter:   Enzo Life Sciences
Microcystin-LR
Katalognr. ENZOALX350012C100
Anbieter:  Enzo Life Sciences
CAS Nummer:  
Microcystin-LR
Microcystin-LR
100 µG
CAS Nummer:  
Preis
SFr. 202.00
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents
Inhibitor of PP1 and PP2A
Heptapeptide ester hepatotoxin. Tumor promoter. Equally potent and selective inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). PP2B is less sensitive and PP2C is not inhibited up to 4 µM. Useful for affinity-purification of PP2A. The product is not cell permeable except in liver cells, which appear to have a functional uptake system. Is absorbed by hepatocytes via the multispecific organic anion transporter. Does not induce any effects on mouse skin or human fibroblasts due to cell membranes impermeability. Has no effect on protein kinases. Less toxic than the more hydrophobic analogs microcystin-LY, -LW and -LF. Frequently contaminates fresh-water lakes and ponds. Causes livestock poisonings. Ozonation did lead to complete loss of toxicity and toxins from contaminated samples.
Isolated from Microcystis aeruginosa.
Technische Daten
Technische Daten
  • VE:
    100 µG
  • Cat. No.:
    ENZOALX350012C100
  • Protein/Peptid-Name:
    Microcystin-LR
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.