Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Okadasäure

Okadasäure

1 mg
Catalog # ENZOALX350003M001
Supplier:   Enzo Life Sciences
Okadasäure
Catalog # ENZOALX350003M001
Supplier:  Enzo Life Sciences
CAS Number:  78111-17-8
Okadasäure
Okadasäure

1 mg

CAS Number:  78111-17-8
Price
SFr. 2’820.00
Log in for availability
1 * 1 mg
Product Details & Documents
Synonyms:  9,10-deepithio-9,10-didehydroacanthifolicin, OA
  • CAS Number:78111-17-8
  • Formula:C₄₄H₆₈O₁₃
  • MDL Number:MFCD00083455
  • Molecular Weight:805.02 g/mol
Specifications

Specifications

  • VE:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.