Okadasäure
50 µG
Katalognr. ENZOALX350003C050
Anbieter: Enzo Life Sciences
Direkt zu
Product Details & Documents
Synonyme: 9,10-deepithio-9,10-didehydroacanthifolicin, OA
- CAS Nummer:78111-17-8
- Formel:C₄₄H₆₈O₁₃
- MDL-Nummer:MFCD00083455
- Molekulargewicht:805.02 g/mol
Technische Daten
Technische Daten
- VE:50 µG
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



