Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
DAPT ≥98% (durch NMR)

DAPT ≥98% (durch NMR)

5 mg
Katalognr. ENZOALX270416M005
Anbieter:   Enzo Life Sciences
DAPT ≥98% (durch NMR)
Katalognr. ENZOALX270416M005
Anbieter:  Enzo Life Sciences
CAS Nummer:  208255-80-5
DAPT ≥98% (durch NMR)

5 mg

CAS Nummer:  208255-80-5
Preis
202.00 SFr.
Einloggen für Verfügbarkeit
1 * 5 mg
Product Details & Documents

gamma-Secretase inhibitor

Cell permeable inhibitor of γ-secretase (IC50=115nM for total β-amyloid, IC50=200 nM for β-amyloid 1-42). It reduces Aβ levels in vivo in plasma and cerebrospinal fluid in young and aged Tg2576 mice. It blocks the proteolytic processing of neurotrophin receptor alike death domain protein (NRADD)Does not inhibit presenilinase. Antagonizes Notch signaling through inhibition of Notch processing. DAPT treatment can influence hematopoietic cell fate decisions and enhances neuronal differentiation in embryonic stem cell-derived embryoid bodies independent of sonic hedgehog (shh) signaling.

Synonyme:  N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
  • CAS Nummer:208255-80-5
  • Formel:C₂₃H₂₆F₂N₂O₄
  • MDL-Nummer:MFCD04974585
  • Molekulargewicht:432.47 g/mol
Technische Daten

Technische Daten

  • VE:
    5 mg

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.