POM Balls for SampleJet Caps
Cap sealing balls, yellow
Catalog # BRUKZ147555
Supplier: BRUKER DALTRONIK
Jump to:
Product Details & Documents
Set of 100 POM balls to close samplejet tube caps. Available in different colours.
Specifications
- Bezeichnung:Cap sealing balls, yellow
- Seq:02