BioSample cards
Product Details & Documents
These BioSample cards are designed for microvolume sampling, transport and ambient storage of biological samples. These cards are made of white and pure absorbent fibers and contain no wet-strength additives or chemicals. This feature ensures reproducible collection and minimise possible interferences for reliable analysis of targeted analytes like small molecules, proteins, nucleic acids or vitamins.
- Reliable and reproducible sample collection
- Even and uniform sample distribution
- Customisable design and printing
- Rigorous quality control
- Each card has five/eight spots
General applications include protein-based analysis, lipid-based analysis, genomics, human identification and forensics. Indicated cards can also be used for HIV viral load testing and infectious disease screening (including Hepatitis B, C and Syphilis).
Zertifizierung: Manufacturing according to ISO 13485:2016 and ISO 9001:2015 standards.
In Vitro Diagnostic medical device in compliance with the Directive 98/79/EC.
Bestellinformationen: Drying racks are available to facilitate drying of the cards at ambient temperature.
Achtung! Bitte beachten: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904
Dokumente
Technische Daten
- Volumen:—
- Bezeichnung:BioSample TFN cards A4 without spots
- Seq:08