Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Ammonia solution 27 - 30% FCC

Ammonia solution 27 - 30% FCC

−, 2,5 L
Catalog # SPCMA1293-2.5LTPV
Corrosive
Irritant
Supplier:   Spectrum Chemical
Ammonia solution 27 - 30% FCC
Catalog # SPCMA1293-2.5LTPV
Supplier:  Spectrum Chemical
CAS Number:  1336-21-6
Ammonia solution 27 - 30% FCC

−, 2,5 L

CAS Number:  1336-21-6
Price
SFr. 217.00
Log in for availability
1 * 2,5 L
Product Details & Documents

Ammonia Solution, FCC is a solution of Ammonia in water. It has many uses, but in food production it is used primarily as a leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Corrosion;Exclamation mark
UN2672

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0,900
  • Storage Temperature:Raumtemperatur
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • Assay (NH₃, By Wt.):
    27.0 - 30.0%
  • Lead (Pb):
    max. 0.5 mg/kg
  • Nonvolatile Residue:
    max. 0.02 %
  • Readily Oxidizable Substances:
    To Pass Test
  • Identification:
    To Pass Test

Specifications

  • Packung:
  • VE:
    2,5 L

Specifications