Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Sucralose NF

Sucralose NF

Poly pail, 25 kg
Katalognr. SPCMS1416-25KGBL
Anbieter:   Spectrum Chemical
Sucralose NF
Katalognr. SPCMS1416-25KGBL
Anbieter:  Spectrum Chemical
CAS Nummer:  56038-13-2
Sucralose NF

Poly pail, 25 kg

CAS Nummer:  56038-13-2
Preis
SFr. 9’790.00
Einloggen für Verfügbarkeit
1 * 25 kg
Product Details & Documents

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Synonyme:  Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside
  • CAS Nummer:56038-13-2
  • Formel:C₁₂H₁₉Cl₃O₈
  • MDL-Nummer:MFCD03648615
  • Molekulargewicht:397.63 g/mol
  • Schmelzender Platingehalt:130 °C
  • Lagertemperatur:Raumtemperatur

Documents

Technische Daten

Ergebnisse des Spezifikationstests

  • APPEARANCE:
    CONFORM
  • ASSAY (ANHYDROUS BASIS):
    98.0 - 102.0 %
  • SPECIFIC ROTATION:
    +84.0° to +87.5°
  • WATER DETERMINATION:
    Max. 2.0 %
  • RESIDUE ON IGNITION:
    Max. 0.7 %
  • CADMIUM (Cd):
    AS REPORTED
  • LEAD (Pb):
    AS REPORTED
  • ARSENIC (As):
    AS REPORTED
  • MERCURY (Hg):
    AS REPORTED
  • HYDROLYSIS PRODUCTS:
    Max. 0.1 %
  • METHANOL:
    Max. 0.1 %
  • RELATED COMPOUNDS:
    Max. 0.5 %
  • IDENTIFICATION A (FTIR):
    SPECTRUM MATCHES REFERENCE
  • IDENTIFICATION (B):
    RETENTION TIME CORRESPONDS TO STANDARD
  • IDENTIFICATION (C):
    Rf VALUE CORRESPONDS TO STANDARD
  • RESIDUAL SOLVENTS:
    AS REPORTED

Technische Daten

  • Packung:
    Poly pail
  • VE:
    25 kg

Technische Daten