Leptomycin B
Bottle, 5 mg
Katalognr. APOSBIL2101-5MG
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Potent inhibitor of nuclear export.
Supplied as a 1mM solution in abs. ethanol
Synonyme: (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
- CAS Nummer:87081-35-4
- Formel:C₃₃H₄₈O₆
- MDL-Nummer:MFCD06795848
- Molekulargewicht:540.73 g/mol
- Blitz Pt:11,1 °C
- Lagertemperatur:Gefrierschrank
- UN:1170
- ADR:3,II
Technische Daten
Technische Daten
- Packung:Bottle
- VE:5 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



